Stirring shafts, two paddles, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C380-02 C380-10 C380-16 C380-08 C380-04 C380-18 C380-14 C380-12
BOHLC380-02EA 489 EUR
BOHLC380-02 BOHLC380-10 BOHLC380-16 BOHLC380-08 BOHLC380-04 BOHLC380-18 BOHLC380-14 BOHLC380-12
Stirring shafts, two paddles, BOLA
Roerders Roerassen
Stirrer shafts made of PTFE-jacketed stainless steel and the paddles made of PTFE.
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C
Two paddles arranged crosswise at 90°. Upper paddle is fixed by means of clamp screws made of PEEK compound.