Wire baskets, autoclavable, Poxygrid®
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H167660000 H167670000 H167590000 H167650000
BELAH167660000EA 256 EUR
BELAH167660000 BELAH167670000 BELAH167590000 BELAH167650000
Wire baskets, autoclavable, Poxygrid®
Manden Draadmanden
These epoxy-coated, steel wire baskets are suitable for transporting and batch dipping glassware.
  • Resistant to rust, corrosion, solvents, acids and alkalis
  • Withstands a temperature range of −223 to +121 °C