Water baths, stainless steel for refrigeration
Leverancier: HUBER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2009.0032.00 2010.0026.00 2011.0022.00 2012.0026.00
HUBE2009.0032.00EA 3540 EUR
HUBE2009.0032.00 HUBE2010.0026.00 HUBE2011.0022.00 HUBE2012.0026.00
Water baths, stainless steel for refrigeration
Baden Waterbaden, ongeroerd
The Cooling Bath are water baths with air-cooled refrigerating unit and use natural refrigerants. Housing and tank of stainless steel. The refrigerating unit works continuously.
In combination with an immersion circulator these refrigerated bath systems offers active cooling, in continuous operation over the complete working range.
Opgelet: Cooling capacity at +20; 0; −10; −20; −30 °C.