Magnetic beads, His Mag Sepharose® Ni
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
28-9673-88 28-9673-90 29-1040-65
28-9673-88EA 112 EUR
28-9673-88 28-9673-90 29-1040-65
Magnetic beads, His Mag Sepharose® Ni
Parels Magnetische parels
His Mag Sepharose® Ni are magnetic beads designed for rapid, small-scale purification and screening of histidine-tagged proteins from different sources.
  • Easy-to-use, visible magnetic beads ensures reliable collection of bound histidine-tagged proteins in purification procedures
  • High purity and yield
  • Easy parallel screening with high repeatability
  • High density of beads ensures rapid capture from magnetic devices
  • Scalable-simple capture of target proteins from small µl to high ml sample volumes
  • Storage condition: Room temperature
Lower concentrations of histidine-tagged proteins are commonly used to wash resins prior to elution. Mag Sepharose® format has excellent properties for small-scale experiments.