Bottles, wide neck, with screw cap
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0301-0250 0301-0100 0301-0500 0301-1000
BURK0301-0250EA 2.2 EUR
BURK0301-0250 BURK0301-0100 BURK0301-0500 BURK0301-1000
Bottles, wide neck, with screw cap
Flessen Kunststof flessen
HDPE, translucent, with PP screw cap.
  • Embossed contents scale
  • Absolutely leakproof
  • Suitable for use with foodstuffs
Bottle for universal use. The medium-sized bottle opening ensures good filling and safe pouring.