WHEATON® E-Z Ex-Traction™ High Recovery Vials
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
W224613 W224618 W224617 W224630 W224634 W224601 W224636 W224613L
WHEAW224613EA 1410 EUR
WHEAW224613 WHEAW224618 WHEAW224617 WHEAW224630 WHEAW224634 WHEAW224601 WHEAW224636 WHEAW224613L
WHEATON® E-Z Ex-Traction™ High Recovery Vials
flesjes
Precision-manufactured from low extractable borosilicate glass, these vials are ideal for compound biological sample applications. Vials conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
  • Conical well ensures maximum sample recovery
  • Quality engineered for automated sample handling systems