Crimpers, E-Z Crimper™, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
W225302 W225300 W225301 W225303
WHEAW225302EA 801 EUR
WHEAW225302 WHEAW225300 WHEAW225301 WHEAW225303
Crimpers, E-Z Crimper™, WHEATON®
Capsuleer- en decapsuleertangen
The E-Z Crimper™ crimpers are used to attach aluminium seals to bottles and vials with a crimp or serum finish.
  • Cushioned ergonomic handle reduces hand fatigue
  • Labeled for quick size identification
  • Polished crimping jaws provide consistent sealing
Leveringsinformatie: The 11 mm E-Z Crimper™ is supplied with a plunger insert to give the option of crimping the top of a vial creating an extra tight seal. We recommend this only for standard opening vials.
Order Now
Specificaties
Environmentally Preferable