Conductors for magnetic stirrer heads, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C463-29 C463-45 C463-19
BOHLC463-29EA 849 EUR
BOHLC463-29 BOHLC463-45 BOHLC463-19
Conductors for magnetic stirrer heads, BOLA
Roerders Roergereedschap
Conductor made of borosilicate glass with or without ground joint.
  • Chemical resistance
  • Temperature resistance: −200 to 250 °C
  • Glued upper bearing is included
Conductor without ground joint is designed to fuse with reactor lid.