Scintillation vials, Sampule®, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
986492 986645
WHEA986492EA 884 EUR
WHEA986492 548-0258
Scintillation vials, Sampule®, WHEATON®
flesjes
Glass vials made from 180 low potassium borosilicate glass that conforms to ASTM E 438 Type 1, Class A and USP Type I requirements.
  • Background counts remain low and ultraviolet transmission high
HDPE vials are the economical choice for scintillation counting.
Verpakking: Caps are packaged separately.