Reusable coliwasa samplers, Wheaton®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
885252 885250
WHEA885252EA 911 EUR
WHEA885252 WHEA885250
Reusable coliwasa samplers, Wheaton®
Monsternemers Monsternemers voor vloeistoffen
Uniquely designed top spin bar and bottom valve plate mechanism provides a positive seal for sample retention along with greater control for releasing the sample.
  • Modular design has added flexibility by allowing the connection of several sections to form a larger sampling unit
  • Spin open-top locking handle, insert slowly into drum to collect liquid column, pull top handle to seat bottom valve plate and spin top handle closed, trapping column in sampler