Vials with inserts, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
225190 225195 225200 225205
WHEA225190EA 453 EUR
WHEA225190 WHEA225195 WHEA225200 WHEA225205
Vials with inserts, WHEATON®
flesjes
These plastic autosampler vials combine thermoplastic polymer outer vial shells with type I borosilicate glass inserts.
  • 100 µl glass insert
  • Available with snap/crimp or screw top finish
  • Conical interior ensures maximum retrieval of contents without the hassle of removable inserts
The thermoplastic polymer provides excellent dimensional tolerance reliability for capping and instrumentation use and safety from breakage.
Order Now
Specificaties
Environmentally Preferable