Biopsieverwerkings-/inbedcassettes, Swingsette®, M518
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M518-10 M518-11 M518-12 M518-2 M518-3 M518-4 M518-5 M518-6 M518-8 M518-9 M518-7
720-1670EA 151 EUR
720-1670 720-1671 720-1672 720-1676 720-1677 720-1678 720-1679 720-1680 720-1682 720-1683 720-1681
Biopsieverwerkings-/inbedcassettes, Swingsette®, M518
Histologie-cassettes Cassettes voor biopsiebehandeling
Ideal for cassette-labelling instruments since the covers are packed separately in the case. These biopsy cassettes, made of acetal polymer, are similar to the M517 series but are especially designed to hold biopsy specimens during the processing/embedding process as well as in storage cabinets.
  • Moulded from a special high density polymer, they keep specimens safely submerged in liquid
  • Totally resistant to the chemical action of histological solvents
  • The efficient flow-through slots maximise fluid exchange and ensure proper drainage
  • The special hinge make it easy to attach lid to base
This new model differs by the special hinge that holds the base and cover together. This hinge allows cassettes to be opened and closed as often as necessary. The cover can be removed and re-inserted easily without danger of specimen loss. Three frosted writing surfaces are made available, including the anterior one at a 45° angle. Available in several colours.
Verpakking: Elke doos bevat twee verdeeldozen met 500 cassettes en één doos met 1000 afdekkingen.