WHEATON® Vials with Inserts
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
225180 225185 W225186
WHEA225180EA 474 EUR
WHEA225180 WHEA225185 WHEAW225186
WHEATON® Vials with Inserts
flesjes
These vials are made of durable PP and can be used in standard gas and liquid chromatography autosamplers.
  • Lightweight and economical
  • Available with snap/crimp or screw top finish
  • Conical interior ensures maximum retrieval of contents without the hassle of using removable inserts
Verpakking: 100 vials per shelf pack or 1000 vials per case.