Eco stopcock plugs, with bore and taper 1:10
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J1220-06 J1220-15 J1220-05 J1220-03 J1220-10 J1220-20 J1220-25 J1220-30 J1235-15 J1220-40 J1220-35 J1220-01 J1255-10
BOHLJ1220-06EA 5.5 EUR
BOHLJ1220-06 BOHLJ1220-15 BOHLJ1220-05 BOHLJ1220-03 BOHLJ1220-10 BOHLJ1220-20 BOHLJ1220-25 BOHLJ1220-30 BOHLJ1235-15 BOHLJ1220-40 BOHLJ1220-35 BOHLJ1220-01 BOHLJ1255-10
Eco stopcock plugs, with bore and taper 1:10
Afsluitkranen
PTFE, white.
  • Very good chemical resistance
  • Very good temperature resistance