PerfeCTa® MultiPlex qPCR SuperMix, low ROX
Leverancier: Quantabio
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
95108-01K 95108-200
733-2086EA 3880 EUR
733-2086 733-2088
PerfeCTa® MultiPlex qPCR SuperMix, low ROX
Reagentia voor nucleïnezuren qPCR/RT-qPCR Enzymes and Kits
PerfeCTa® Multiplex qPCR SuperMix, low ROX is a 2X concentrated, ready to use reaction cocktail that contains all components, except primers, probe(s), and template for real time qPCR on Applied Biosystems 7500 or Stratagene MX series of real time PCR systems.
PerfeCTa® Multiplex qPCR SuperMix, low ROX delivers dynamic range and sensitivity to multiplexed qPCR that is comparable to that for singleplex qPCR probe assays without the need for limiting or variable primer concentrations.
Leveringsinformatie: Each kit contains 2X reaction buffer containing optimised concentrations of MgCl₂, dNTPs (dATP, dCTP, dGTP, dTTP), AccuStart™ Taq DNA polymerase, ROX reference dye (for 580 to 585 nm excitation), and stabilisers.