Arsine generator apparatus, Clear-Seal™, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
180020 180021 180022
WHEA180020EA 284 EUR
WHEA180020 WHEA180021 WHEA180022
Arsine generator apparatus, Clear-Seal™, WHEATON®
Destructiesystemen Detectieapparaat voor sporenelementen
Engineered for arsine analysis using the silver diethyldithiocarbamate (SDDC) colorimetric method.
Manufactured from 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Certificaten: Meets ASTM / USP and EPA specifications.