Arsine generator apparatus, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
180034 180030 970440
WHEA180034EA 161 EUR
WHEA180034 WHEA180030 WHEA970440
Arsine generator apparatus, WHEATON®
Destructiesystemen Detectieapparaat voor sporenelementen
Designed for arsenic analysis using silver diethyldithiocarbamate (SDDC) colorimetric method.
  • Assembled apparatus is freestanding and easy to use
  • Design eliminates the ball and socket joint connecting the scrubber and absorber components
Manufactured from 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.