Vials, Uni-Dose®, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
226732 226733 226773
WHEA226732EA 584 EUR
WHEA226732 WHEA226733 WHEA226773
Vials, Uni-Dose®, WHEATON®
flesjes
These vials are manufactured from 900 soda-lime amber glass that conforms to USP Type III requirements.
  • Available in amber glass or amber PET (Polyethylene Terephthalate)
  • Use with tamper evident safety cap
  • Large labeling surface for easy product identification
  • Mouth OD 28,5 mm
Order Now
Specificaties
Environmentally Preferable