Closures and pour rings for media bottles, Lab 45™, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
240726 240746 240756 240736 240760 240750 240740 240726-03
WHEA240726EA 57.3 EUR
WHEA240726 WHEA240746 WHEA240756 WHEA240736 WHEA240760 WHEA240750 WHEA240740 WHEA240726-03
Closures and pour rings for media bottles, Lab 45™, WHEATON®
Sluitingen Flesdoppen
Made of PP.
  • 45 mm screw thread
  • Inner sealing ring
  • Available with liner and center hole
  • Autoclavable