VWR®, Toebehoren voor blokkachels analoog/digitaal
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
460-3240EA 174 EUR
460-3240 460-3241 460-3242 460-3289
VWR®, Toebehoren voor blokkachels analoog/digitaal
Blokverwarmers Analoge blokverwarmers
Plexiglas® covers with 2 open sides reduce airflow, providing additional temperature stability in low temperature applications.