Filter candles, cylindrical, VitraPOR®
Leverancier: ROBU GLASFILTER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
18410 18411 18412 18413 18414 18350 18351 18352 18353 18354
ROBU18410EA 189 EUR
ROBU18410 ROBU18411 ROBU18412 ROBU18413 ROBU18414 ROBU18350 ROBU18351 ROBU18352 ROBU18353 ROBU18354
Filter candles, cylindrical, VitraPOR®
Filters Filterbuizen
These cylindrical filter candles are available with or without tube.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
Made of borosilicate glass 3.3.