Cryogenic vial boxes, Cryule®, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
651490 651492
WHEA651490EA 288 EUR
WHEA651490 WHEA651492
Cryogenic vial boxes, Cryule®, WHEATON®
Dozen Dozen voor cryobuisjes
These cryoboxes are made of high quality chip-board and spray coated with polyethylene.
  • Stores 1 and 2 ml cryogenic vials
  • Holds 81 freestanding or round bottom vials
Suitable for use with vapor phase of liquid nitrogen.