Kwartscontroleplaat
Leverancier: BELLINGHAM STANLEY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
34-21 34-20 34-241 34-22 90-803
635-0023EA 1893 EUR
635-0023 635-0022 635-0666 635-0405 BELL90-803
Kwartscontroleplaat
Standaarden Standaarden voor brekingsindexbepaling
Quartz control plates for the calibration and verification of polarimeters. of optical and digital polarimeters and saccharimeters.
  • Inert quartz with relatively low temperature coefficient, easy to handle, and infinite shelf life
  • Available in three optical rotation values
  • Three decimal place uncertainty
The optional temperature saddles may used as an interface between the quartz plate and instrument temperature sensor to provide fine compensation.
Certificaten: Traceable to PTB. UKAS-MRA calibration in accordance with ISO 17025:2005 between 325-880nm available.
Bestelinformatie: Sadlles can be ordered in option.
Leveringsinformatie: Supplied with a certificate of verification in a plastic storage case.