Gooch Filters with Fused Filter Disc and Standard Joint 39/32 or 34/35
Leverancier: ROBU GLASFILTER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45300 45301 45302 45303 45304 45305 45500 45501 45502 45503 45505 45504
ROBU45300EA 81.2 EUR
ROBU45300 ROBU45301 ROBU45302 ROBU45303 ROBU45304 ROBU45305 ROBU45500 ROBU45501 ROBU45502 ROBU45503 ROBU45505 ROBU45504
Gooch Filters with Fused Filter Disc and Standard Joint 39/32 or 34/35
Trechters Filtertrechters Filtertrechters
ROBU® Gooch filter with fused filter disc is ideal for gravimetric analysis and residue collection, combining ROBU® construction with high chemical and thermal resistance, available with integrated VitraPOR® filter for efficient filtration.
  • Robust design
  • Made from high-quality borosilicate glass
  • Fused VitraPOR® filter disc ensures consistent and reliable filtration
  • Chemical resistance and thermal stability
  • Withstands aggressive substances and solvents
  • Suitable for high-temperature applications
  • Ideal for gravimetric and analytical procedures
  • Material: Borosilicate glass 3.3, plastic, silicone
ROBU Gooch filter with fused filter disc is specifically designed for applications requiring precise and consistent filtration, such as gravimetric analysis and the collection of precipitates. Manufactured from durable borosilicate glass, it features a permanently fused VitraPOR filter disc that delivers consistent pore structure and performance.
This construction eliminates the risk of filter displacement and ensures long-term durability, even under harsh chemical and thermal conditions. Its compatibility with standard laboratory setups makes it a reliable and efficient solution for filtration tasks where accuracy, chemical resistance, and robustness are essential.
Certificaten:
In compliance with ISO 4796-80 and ASTM E128-99 standards.