Bottles, wide neck, graduated, Valumetric®, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
W216989 W216975 W216971 W216967 W216968 W216969 W216974 W216972 W216973
215-4467EA 584 EUR
215-4467 WHEAW216975 215-4464 215-4461 215-4462 215-4463 215-4466 WHEAW216972 215-4465
Bottles, wide neck, graduated, Valumetric®, WHEATON®
Flessen Glazen flessen
Soda-lime glass, class III, clear.
  • Ideal for general storage
  • Easy removal of contents
  • Hazardous communication label included with each case of bottles allows for content ID and safety notification
  • Tamper-evident seal included with each case of bottles keeps bottle sealed which protects contents from accidental misuse