Aluminium seals for headspace vials, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
W224216 W224215 W224217
WHEAW224216EA 317 EUR
WHEAW224216 WHEAW224215 WHEAW224217
Aluminium seals for headspace vials, WHEATON®
Sluitingen Sluitingen voor chromatografievials
These aluminum seals are crimp closures with bridges and scorelines allowing the internal pressure to be released when 0,3 ±0,5 bar has been exceeded.
Bestelinformatie: Seals should be ordered separately or together with vials in convenience packs.