Culture tubes with screw cap, C-TUBE®, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
358646 358650 358647 358606 358607
391-0197EA 462 EUR
391-0197 391-0198 WHEA358647 WHEA358606 WHEA358607
Culture tubes with screw cap, C-TUBE®, WHEATON®
Buisjes Cultuurbuisjes
Manufactured from 51 expansion borosilicate glass that conforms to ASTM Type I, Class B and USP Type I requirements.
  • For use as slant or shake cultures
  • Autoclavable
  • Very good chemical resistance
The tubes can be reused many times. The caps have a white 14B styrene-butadiene rubber liner or a PTFE faced 14B styrene-butadiene rubber liner.