Eppendorf Easypet® 3 Accessories and Spare Parts
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4420803.007 4420810.003 4421601.009 4430603.006 4430604.002 4430605.009 4430606.005 4430601.003
612-7037EA 92.7 EUR
612-7037 613-4289 613-0210 EPPE4430603.006 EPPE4430604.002 EPPE4430605.009 612-4899 EPPE4430601.003
Eppendorf Easypet® 3 Accessories and Spare Parts
Pipetten Pipetvullers
Accessories and spare parts for the Eppendorf Easypet® 3 electronic pipette controller.
The controller gives you precision and complete speed control, making it ideal for serial dispensing and resuspending bacterial or cell pellets. Easypet 3 can be used with all types of serological and volumetric pipettes from volumes of 0,1 -  100 ml.