Crimp neck vials, E-Z Vial®, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
W225175-0303 225175 225173
WHEAW225175-0303EA 135 EUR
WHEAW225175-0303 WHEA225175 WHEA225173
Crimp neck vials, E-Z Vial®, WHEATON®
flesjes
E-Z Vials are manufactured from Type I borosilicate glass and feature a 40% larger neck opening than standard opening vials.
  • Use with 11 mm aluminum seals
  • Limited volume insert
The larger opening improves sample accessibility while reducing auto sampler needle damage.
Verpakking: Packaged in shrink-wrapped trays of 100 for convenience and cleanliness.