Geleidbaarheidsstandaarden, Certipur®, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.01203.0500 1.01254.0500 1.01255.0500 1.01553.0001 1.01554.0001 1.01557.0500 1.01586.0001 1.01811.0105
1.01203.0500EA 177.47 EUR
1.01203.0500 1.01254.0500 1.01255.0500 1.01553.0001 1.01554.0001 1.01557.0500 1.01586.0001 1.01811.0105
Geleidbaarheidsstandaarden, Certipur®, Supelco®
Standaarden Geleidbaarheidsstandaarden
These conductivity sachets are convenient, easy to use and ensure accurate and reliable results both in the field or in the laboratory. The 30 ml sachets always provide a fresh solution preventing contamination by microorganisms, CO₂ or other substances. A conductivity sensor can be simply inserted into the sachet.
  • Traceerbaar naar PTB van NIST
  • Getest op basis van primaire referentiestandaarden
Certipur conductivity standards are also available in 500 ml packs for the user with higher volume requirements.