Reagent sets for measurement of nitrogen and nitrates - Test 'N Tube method
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2605345 2606945 2672245 2604945 2604545 2714100
705-0161EA 241 EUR
705-0161 705-0162 705-0175 705-0160 705-0159 705-0181
Reagent sets for measurement of nitrogen and nitrates - Test 'N Tube method
Milieutestkits
Suitable for measurement with selected Hach photometers.
  • Convenient testing in a tube, with minimal handling of chemicals
  • Digestion (where required) is performed in the same tube as the test
Opgelet: The reagent sets may contain materials which are hazardous. Please read the usage instructions and MSDS for details of handling and disposal.