Reagent set for measurement of phosphates and ortho-phosphates - Test 'N Tube method
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2742545 2742645 2742745 2767245
705-0182EA 134 EUR
705-0182 705-0183 705-0184 705-0194
Reagent set for measurement of phosphates and ortho-phosphates - Test 'N Tube method
Milieutestkits
Suitable for measurement with selected Hach photometers which are suitable for 16 mm tubes.
  • Convenient testing in tube, with minimal handling of chemicals
  • Digestion (where required) is performed in the same tube as the test
Opgelet: The reagent sets may contain materials which are hazardous. Please read the usage instructions and MSDS for details of handling and disposal.