Gas washing bottles, VitraPOR®
Leverancier: ROBU
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VitraPOR®
ROBU37350EA 166 EUR
ROBU37350 ROBU37352 201-1465
Gas washing bottles, VitraPOR®
Flessen Gaswasflessen
Borosilicat glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With sintered disc and standard joint cap.