Office furniture
Leverancier: HAMMERBACHER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
V6000/N V6100/N/N V6770/N/N V7600/N/N V7600/N/S VSC50/N VSET50/N/A
132-0765EA 0 EUR
132-0765 132-0761 132-0762 132-0763 132-0764 132-0760 132-0766
Office furniture
Meubilair Opslagkasten
Office cupboard made from a walnut. The main advantage of this line is that the different models can be assembled according to the user's needs or used separately
  • Ergonomic and convinient
  • Adjustable height
  • Shelves: 19 mm thick, with a load capacity of 20 kg
Leveringsinformatie: The models are not assembled for delivery.