Round microcentrifuge floating bubble racks, Bel-Art ProCulture, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F188750400 F188751000 F188751600
211-7148EA 33.5 EUR
211-7148 211-7149 BELAF188751600
Round microcentrifuge floating bubble racks, Bel-Art ProCulture, SP Bel-Art
Rekken Tube Racks
Made especially for use with beakers and waterbaths, these space-saving racks are ideal for small quantities of tubes and for use in cell culture laboratories.
  • Buoyant TPX® brand polymethylpentene with tiny bubbles throughout for additional buoyancy
  • Insert tubes from the top and easily remove them by pressing the rack down on the bench
  • Withstands boiling for 30 minutes at 100 °C (212 °F)
  • Steam autoclavable at 121 °C (250 °F)
  • Leg length: 19 mm (³/₄"); thickness: 6,4 mm (¼")
Perfect for incubating microtubes in a beaker of water.
Order Now
Specificaties
Environmentally Preferable