Wireless monitoring systems, Smart-Vue
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SV206-101 SV104-502 SV200-101 SV200-201 SV201-101 SV201-201 SV202-101 SV202-201 SV203-101 SV203-201 SV204-201 SV100-501 SV209-101 SV509-500 SV500-501 SV212-501 SV205-101 SV209-201 SV208-101 SV207-201 SV207-101 SV206-201 SV103-503 SV106-503-LSB SV103-511-LSB SV103-512-LSB SV103-513-LSB SV103-514-LSB SV603-500-LSB SV604-500-LSB SV605-500-LSB SV606-500-LSB SV607-500-LSB SV608-500-LSB SV609-500-LSB SV610-500-LSB SV611-500-LSB SV612-500-LSB
620-1895EA 515 EUR
620-1895 620-1879 620-1890 THBRSV200-201 620-1891 THBRSV201-201 620-1892 THBRSV202-201 620-1893 THBRSV203-201 THBRSV204-201 620-1887 THBRSV209-101 620-1898 620-1897 THBRSV212-501 620-1894 THBRSV209-201 620-1896 THBRSV207-201 620-1876 THBRSV206-201 620-1877 THBRSV106-503-LSB THBRSV103-511-LSB THBRSV103-512-LSB THBRSV103-513-LSB THBRSV103-514-LSB THBRSV603-500-LSB THBRSV604-500-LSB THBRSV605-500-LSB THBRSV606-500-LSB THBRSV607-500-LSB THBRSV608-500-LSB THBRSV609-500-LSB THBRSV610-500-LSB THBRSV611-500-LSB THBRSV612-500-LSB
Wireless monitoring systems, Smart-Vue
Bewakingssystemen Bewakingssystemen voor CO2 en temperatuur
Ambient measurements or monitoring of laboratory equipment.
  • Easy to read display of monitored setpoints directly on module
  • Wireless data transmission (868 MHz): No wiring or rebuilding required for installation
  • Fully integrated open software: Data can easily be exported to Excel, Word or PDF
  • Safety signal transfer: Automatic signal recognition makes it possible to use each module as repeater
  • Accessible anywhere via the internet with Smart-Vue client software
These instruments safeguard the integrity of precious samples by continuously monitoring and reporting critical conditions, such as temperature, relative humidity, CO₂ percentage and differential pressure. They can be used in highly controlled environments, pharmaceutical production, biotechnology, IVF clinics, clinical studies and diagnostic laboratories, as well as in storage areas. If high/low setpoints are reached, notifications are immediately sent to the user by means of the chosen alert method (visual siren, fax, telephone, email or text messaging).
Traceability to 21 CFR part 11
Certificaten: Traceability to 21 CFR part 11
Bestelinformatie: More modules for monitoring CO₂, pressure, mA analogue output signals and dry contact inputs are available. Please contact your local Avantor representative for further information on models and additional accessories.
Leveringsinformatie: Supplied with a module, sensor, 3,6 V lithium battery, mounting holder with Velcro, magnet & hardware, CD ROM with software drivers and user manuals. Repeater (optional), receiver (required) and alarm systems need to be ordered separately.
Opgelet: * Alarm system equipped with integrated 3,6 V lithium battery for memory back-up, prevents loss of data when power fails.