Multi-tube racks, No-Wire™, SP Bel-Art
Leverancier: Bel-Art
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
No-Wire™
212-8710EA 102 EUR
212-8710 212-8711
Multi-tube racks, No-Wire™, SP Bel-Art
Rekken Tube Racks
Racks, made of PP, white. Choose from two space saving rack configurations, both styles accommodate round and conical bottom tubes.
  • Very good chemical and temperature resistance
  • Shipped flat; easy assembly
  • Steam autoclavable at 121 °C