Pressure compensation sets and membranes, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N1698-18 N1699-18 N1699-45 N1699-32 N1699-25 N1699-14 N1698-45 N1698-32 N1698-25 N1698-14
BOHLN1698-18EA 10.7 EUR
BOHLN1698-18 BOHLN1699-18 BOHLN1699-45 BOHLN1699-32 BOHLN1699-25 BOHLN1699-14 BOHLN1698-45 BOHLN1698-32 BOHLN1698-25 BOHLN1698-14
Pressure compensation sets and membranes, BOLA
Ventielen Overdrukventielen
Pressure compensation sets consisting of screw cap made of PPS for GL thread, PTFE/silicone gasket and exchangeable filtering membrane made of PTFE. Filtering membranes (2 µm) made of PTFE.
  • Temperature resistance: from −200 to +230 °C
  • Universal chemical resistance
  • Vacuum suitable
  • Autoclavable: 121 °C
For pressure compensation during filling or drawing of liquids. Prevention of unintentional overpressure or vacuum in the vessel. Integrated membrane prevents contamination of the product.