Connection bolts with sealing cone, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F830-10 F833-10 F837-10 F836-10 F834-10
BOHLF830-10EA 14 EUR
BOHLF830-10 BOHLF833-10 BOHLF837-10 BOHLF836-10 BOHLF834-10
Connection bolts with sealing cone, BOLA
Bevestigingen Schroeven
PEEK, beige.
  • Temperature resistance: from −50 to +250 °C
  • Very good chemical resistance
  • Pressure: 450 bar
  • FDA conform
Connection bolt and sealing cone made of PEEK, suitable for female thread UNF 10-32G.