Pastette® transfer pipettes, standard
Leverancier: Alpha Laboratories
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LW4237 LW4021
ALPCLW4237EA 153 EUR
ALPCLW4237 ALPCLW4021
Pastette® transfer pipettes, standard
Pipetten Pasteur-/transferpipetten
Use these disposable Pastettes instead of a Pasteur pipette for liquid transfers.
Ideal for accessing small diameter tubes such as cuvettes and analyser cups. The stems can be heat-sealed to create a deep freeze, transport or storage container.
Designed to replace glass Pasteur pipettes, Pastette (transfer pipette) s are unbreakable and don't require a separate bulb. and, as they are manufactured from soft low density polyethylene (LDPE), can be recycled if used with non hazardous samples.
Verpakking: Pack of 500 individually wrapped.