Sulphur standard solution, 1000 mg/L S in water (from (NH₄)₂SO₄), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13842.AD 13842.AE 13842.AP
13842.ADEA 55.5 EUR
13842.AD 13842.AE 13842.AP
Sulphur standard solution, 1000 mg/L S in water (from (NH₄)₂SO₄), standard for ICP, Specpure®
Standaarden Kationische standaarden Zwavel standaarden
Molecular Formula: (NH₄)₂SO₄ in H₂O
Physical Form: Liquid
MDL No.: MFCD00085316
Certificaten: Traceable to NIST