Germanium standard solution, 1000 mg/L Ge in water with hydrofluoric acid (max. 1%) (from (NH?)?GeF?), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42242.AD 42242.AE 42242.AP
42242.ADEA 67.7 EUR
42242.AD 42242.AE 42242.AP
Germanium standard solution, 1000 mg/L Ge in water with hydrofluoric acid (max. 1%) (from (NH?)?GeF?), standard for ICP, Specpure®
Standaarden Kationische standaarden Germanium standaarden
Molecular Formula: (NH₄)₂GeF₆ in H₂O/trace F-
Physical Form: Liquid
MDL No.: MFCD00085310
Certificaten: Traceable to NIST