2-Phenyl-2-propanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: α,α-Dimethylbenzyl alcohol

P0213-25G P0213-500G P0213-100G
TCIAP0213-25GEA 62.7 EUR
TCIAP0213-25G TCIAP0213-500G TCIAP0213-100G
2-Phenyl-2-propanol ≥98.0% (by GC)
2-Phenyl-2-propanol
Formule: C₆H₅C(OH)(CH₃)₂
Kookpunt: 215°C
Smeltpunt: 33…35 °C
Dichtheid: 0,973 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004456
CAS nummer: 617-94-7
EINECS: 210-539-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=838b3aba920a8f277439b6b9f41f8019bd98d0c4&vid=838b3aba920a8f277439b6b9f41f8019bd98d0c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR