Praseodymium standard solution, 1000 mg/L Pr in 5% nitric acid (from Pr₆O₁₁), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88088.AE
88088.AP
88088.AEEA
46.4
EUR
88088.AE
88088.AP
Praseodymium standard solution, 1000 mg/L Pr in 5% nitric acid (from Pr₆O₁₁), standard for AAS, Specpure®
Standaarden
Kationische standaarden
Praseodymium standaarden
Molecular Formula: Pr₆O₁ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011174
Certificaten: Traceable to NIST