Rhodium standard solution, 1000 mg/L Rh in 20% hydrochloric acid (from RhCl₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
35754.AD 35754.AP 35754.AE
35754.ADEA 151 EUR
35754.AD 35754.AP 35754.AE
Rhodium standard solution, 1000 mg/L Rh in 20% hydrochloric acid (from RhCl₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Rodium standaarden
Molecular Formula: RhCl₃ in 20% HCl
Physical Form: Liquid
Odour: Characteristic
UN No.: 1789
MDL No.: MFCD00011201
Certificaten: Traceable to NIST