Rhodium standard solution, 10000 mg/L Rh in 20% hydrochloric acid (from RhCl₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
35763.AD
35763.AE
35763.AP
35763.ADEA
1238
EUR
35763.AD
35763.AE
35763.AP
Rhodium standard solution, 10000 mg/L Rh in 20% hydrochloric acid (from RhCl₃), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Rodium standaarden
Molecular Formula: RhCl₃ in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011201
Certificaten: Traceable to NIST