Ferrocene, high purity
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B25007.18 B25007.30
B25007.18EA 34.2 EUR
B25007.18 B25007.30
Ferrocene, high purity
Ferrocene
Formule: C₁₀H₁₀Fe
Kookpunt: 249 °C (1013 hPa)
Smeltpunt: 172…175 °C
Dichtheid: 1,49 g/cm³ (20 °C)
MDL Nummer: MFCD00001427
CAS nummer: 102-54-5
EINECS: 203-039-3
UN: 1325
ADR: 4.1,II
Merck Index: 13,04070