Platinum standard solution, 10000 mg/L Pt in 20% hydrochloric acid (from Pt), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14397.AD
14397.AE
14397.AP
14397.ADEA
360
EUR
14397.AD
14397.AE
14397.AP
Platinum standard solution, 10000 mg/L Pt in 20% hydrochloric acid (from Pt), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Platina standaarden
Molecular Formula: Pt in 20% HCl
Physical Form: Liquid
Odour: Characteristic
UN No.: 1789
MDL No.: MFCD00011179
Certificaten: Traceable to NIST