Platinum standard solution, 1000 mg/L Pt in 20% hydrochloric acid (from Pt), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13827.AD 13827.AE 13827.AP
13827.ADEA 105 EUR
13827.AD 13827.AE 13827.AP
Platinum standard solution, 1000 mg/L Pt in 20% hydrochloric acid (from Pt), standard for ICP, Specpure®
Standaarden Kationische standaarden Platina standaarden
Molecular Formula: Pt in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011179
Certificaten: Traceable to NIST
Order Now
Specificaties
Environmentally Preferable
MDL No. MFCD00011179