Platinum standard solution, 1000 mg/L Pt in 20% hydrochloric acid (from Pt), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88086.AE 88086.AP
88086.AEEA 85.2 EUR
88086.AE 88086.AP
Platinum standard solution, 1000 mg/L Pt in 20% hydrochloric acid (from Pt), standard for AAS, Specpure®
Standaarden Kationische standaarden Platina standaarden
Molecular Formula: Pt in 20% HCl
Physical Form: Liquid
Odour: Characteristic
UN No.: 1789
MDL No.: MFCD00011179
Certificaten: Traceable to NIST